missing translation for 'onlineSavingsMsg'
Learn More
3-Amino-4,4,4-trifluorobutyric acid hydrochloride, 95%, Thermo Scientific™ Product Code.: 15480847

3-Amino-4,4,4-trifluorobutyric acid hydrochloride, 95%, Thermo Scientific™

Product Code. 15480847
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15480847

Brand: Thermo Scientific Alfa Aesar L18336.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91291-66-6
Molecular Formula C4H7ClF3NO2
Molecular Weight (g/mol) 193.55
MDL Number MFCD00181951
InChI Key MRMARVCQZOYQPI-UHFFFAOYNA-N
Synonym 3-amino-4,4,4-trifluorobutanoic acid hydrochloride, 3-amino-4,4,4-trifluorobutyric acid hydrochloride
PubChem CID 14442472
IUPAC Name 3-amino-4,4,4-trifluorobutanoic acid;hydrochloride
SMILES Cl.NC(CC(O)=O)C(F)(F)F

Specifications

Melting Point 197°C to 199°C
Quantity 250 mg
Formula Weight 193.55
Percent Purity 95%
Chemical Name or Material 3-Amino-4,4,4-trifluorobutyric acid hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Amino-4,4,4-trifluorobutyric acid hydrochloride, 95%, Thermo Scientific™ >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado