missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-hydroxypyridine, 98%, Thermo Scientific Chemicals

329.12 NOK - 5430.00 NOK

Chemical Identifiers

CAS 33630-99-8
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD03411556
InChI Key VTSFNCCQCOEPKF-UHFFFAOYSA-N
Synonym 3-amino-2-hydroxypyridine, 3-aminopyridin-2-ol, 3-amino-2-pyridinol, 3-aminopyridin-2 1h-one, 2 1h-pyridinone, 3-amino, 3-amino-2-pyridone, 3-amino-pyridin-2-ol, 2-pyridinol, 3-amino, 2-hydroxy-3-aminopyridine, 3-aminohydropyridin-2-one
PubChem CID 322353
SMILES NC1=CC=CNC1=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15421709
SDS View Documents
Thermo Scientific Alfa Aesar
H64480.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15401709
SDS View Documents
Thermo Scientific Alfa Aesar
H64480.03
1 g
1235.00 NOK
1g
Please to purchase this item. Need a web account? Register with us today!
15411709
SDS View Documents
Thermo Scientific Alfa Aesar
H64480.06
5 g
5430.00 NOK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as pharmaceutical intermediate. It is involved in synthesis of functionalized pyrido[4,3-b][1,4]oxazine and imidazo[1,2-a]pyridine derivatives. The use of ethyl 2-chloro-3-oxopropanoate with 2-amino-3-hydroxypyridine or 3-amino-4-hydroxypyridine led, respectively, to imidazo[1,2-a]pyridine derivatives or ethyl pyrido[4,3-b][1,4] oxazine-2-carboxylate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. It is involved in synthesis of functionalized pyrido[4,3-b][1,4]oxazine and imidazo[1,2-a]pyridine derivatives. The use of ethyl 2-chloro-3-oxopropanoate with 2-amino-3-hydroxypyridine or 3-amino-4-hydroxypyridine led, respectively, to imidazo[1,2-a]pyridine derivatives or ethyl pyrido[4,3-b][1,4] oxazine-2-carboxylate.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

missing translation for 'storageNote' : 2° to 8°C

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Amino-2-hydroxypyridine, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.