missing translation for 'onlineSavingsMsg'
Learn More
3,5-Dihydroxybenzaldehyde, 98% Product Code.: 11468246

3,5-Dihydroxybenzaldehyde, 98%

Product Code. 11468246
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11468246

Brand: Thermo Scientific Alfa Aesar A15338.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26153-38-8 | C7H6O3 | 138.12 g/mol

3,5-Dihydroxybenzaldehyde is used as a building block in the synthesis of more complex structures. It is also used in the synthesis of terbutaline (T109750), which is an important bronchodilator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dihydroxybenzaldehyde is used as a building block in the synthesis of more complex structures. It is also used in the synthesis of terbutaline (T109750), which is an important bronchodilator.

Solubility
Soluble in ethanol and diethyl ether.

Notes
Air sensitive. Store in cool place. Incompatible with strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26153-38-8
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00016611
InChI Key HAQLHRYUDBKTJG-UHFFFAOYSA-N
Synonym benzaldehyde, 3,5-dihydroxy, 3,5-dihydroxy benzaldehyde, 5-formylresorcinol, 5-formylbenzene-1,3-diol, pubchem8249, alpha-resorcylic aldehyde, acmc-1cg7h, 3,5-dihydoxybenzal dehyde, 3,5-dihydroxy-benzaldehyde, .alpha.-resorcylic aldehydeShow MoreShow Less
PubChem CID 94365
ChEBI CHEBI:50204
IUPAC Name 3,5-dihydroxybenzaldehyde
SMILES OC1=CC(C=O)=CC(O)=C1

Specifications

Melting Point 158°C to 161°C
Assay Percent Range 98%
Quantity 1 g
Beilstein 1930147
Sensitivity Air Sensitive
Solubility Information Soluble in ethanol and diethyl ether.
Formula Weight 138.12
Percent Purity 98%
Chemical Name or Material 3,5-Dihydroxybenzaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberT21I019 Certificate TypeCertificate of Analysis Date29/07/2022 Product CodeA15338.06,A15338.03,A15338.14
Lot Number10173848 Certificate TypeCertificate of Analysis Date13/07/2022 Product CodeA15338.06,A15338.03,A15338.14
Lot NumberI6508A Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA15338.06,A15338.03,A15338.14
Lot Number10100202 Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA15338.06,A15338.03,A15338.14
Lot NumberI25T003 Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA15338.06,A15338.03,A15338.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,5-Dihydroxybenzaldehyde, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.