missing translation for 'onlineSavingsMsg'
Learn More
3,5-Diaminobenzoic acid, 96%, may contain up to 3% moisture, Thermo Scientific Chemicals Product Code.: 11349529

3,5-Diaminobenzoic acid, 96%, may contain up to 3% moisture, Thermo Scientific Chemicals

Product Code. 11349529
100 g
Click to view available options
Quantity:
50 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11349529

Brand: Thermo Scientific Alfa Aesar A14075.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 535-87-5 | C7H8N2O2 | 152.153 g/mol

3,5-Diaminobenzoic acid is an analog of diatriazoate, which has been shown to have antigenicity properties towards the monoclonal antibodies. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Diaminobenzoic acid is an analog of diatriazoate, which has been shown to have antigenicity properties towards the monoclonal antibodies. It is used as pharmaceutical intermediate.

Solubility
Very soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 535-87-5
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00007807
InChI Key UENRXLSRMCSUSN-UHFFFAOYSA-N
Synonym benzoic acid, 3,5-diamino, 1-carboxy-3,5-diaminobenzene, ccris 2885, 3,5-diamino benzoic acid, 3,5-diaminobenzoicacid, dabz, pubchem21025, 3,5-diaminebenzoicacid, acmc-1awxi, 3,5-diamino-benzoic acid
PubChem CID 12062
IUPAC Name 3,5-diaminobenzoic acid
SMILES C1=C(C=C(C=C1N)N)C(=O)O

Specifications

Melting Point ∼235°C (decomposition)
Flash Point 210°C (410°F)
Quantity 100 g
Beilstein 2086484
Merck Index 14,2978
Solubility Information Very soluble in water.
Formula Weight 152.15
Percent Purity 98%
Assay may contain up to 3% moisture
Chemical Name or Material 3,5-Diaminobenzoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10248684 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog NumberA14075.36,A14075.22,A14075.18
Lot Number10243179 Certificate TypeCertificate of Analysis Date06/09/2022 Catalog NumberA14075.36,A14075.22,A14075.18
Lot Number10236098 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA14075.36,A14075.22,A14075.18
Lot Number10140779 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14075.36,A14075.22,A14075.18
Lot Number10188075 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14075.36,A14075.22,A14075.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,5-Diaminobenzoic acid, 96%, may contain up to 3% moisture, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.