missing translation for 'onlineSavingsMsg'
Learn More
3-[(4-Methylpiperazin-1-yl)methyl]benzonitrile, ≥97%, Thermo Scientific™ Product Code.: 10177892

3-[(4-Methylpiperazin-1-yl)methyl]benzonitrile, ≥97%, Thermo Scientific™

Product Code. 10177892
1 g, Amber Glass Bottle, 1g
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10177892

Brand: Thermo Scientific Maybridge CC45316DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 859850-90-1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C13H17N3
MDL Number MFCD07772861
Quantity 1 g
Synonym 3-4-methylpiperazin-1-yl methyl benzonitrile, 3-4-methyl-piperazin-1-yl-methyl-benzonitrile, 3-4-methyl-piperazin-1-ylmethyl-benzonitrile, 3-4-methylpiperazinyl methyl benzenecarbonitrile, benzonitrile,3-4-methyl-1-piperazinyl methyl
InChI Key NNYDAONDPGWBRI-UHFFFAOYSA-N
SMILES CN1CCN(CC2=CC=CC(=C2)C#N)CC1
IUPAC Name 3-[(4-methylpiperazin-1-yl)methyl]benzonitrile
Molecular Weight (g/mol) 215.30
PubChem CID 7164647
Formula Weight 215.3
Percent Purity 97%
Chemical Name or Material 3-[(4-Methylpiperazin-1-yl)methyl]benzonitrile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-[(4-Methylpiperazin-1-yl)methyl]benzonitrile, ≥97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.