missing translation for 'onlineSavingsMsg'
Learn More
Alfa Aesar™ 3-(4-Methoxyphenyl)-2-phenylpropionic acid Product Code.: 11439320

Alfa Aesar™ 3-(4-Methoxyphenyl)-2-phenylpropionic acid

Product Code. 11439320
250 mg, 250mg
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11439320

Brand: Alfa Aesar™ H50213.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 4314-68-5
Molecular Formula C16H16O3
Molecular Weight (g/mol) 256.301
MDL Number MFCD00046532
InChI Key GIROORGDVPRQFZ-UHFFFAOYSA-N
Synonym 3-4-methoxyphenyl-2-phenylpropanoic acid, 3-4-methoxy-phenyl-2-phenyl-propionic acid, 3-4-methoxyphenyl-2-phenylpropionic acid, chembl83949, acmc-20anjp, benzenepropanoic acid,4-methoxy-a-phenyl, 4-methoxy-alpha-phenylbenzenepropanoic acid, 3-4-methoxyphenyl-2-phenyl-propanoic acid, 3-p-methoxyphenyl-2-phenylpropionic acidShow MoreShow Less
PubChem CID 97739
IUPAC Name 3-(4-methoxyphenyl)-2-phenylpropanoic acid
SMILES COC1=CC=C(C=C1)CC(C2=CC=CC=C2)C(=O)O

Specifications

Quantity 250 mg
Formula Weight 256.3
Chemical Name or Material 3-(4-Methoxyphenyl)-2-phenylpropionic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberK05S058 Certificate TypeCertificate of Analysis Date08/12/2021 Product Code
Lot NumberJ23R056 Certificate TypeCertificate of Analysis Date08/12/2021 Product Code
Lot NumberK16Q018 Certificate TypeCertificate of Analysis Date08/12/2021 Product Code
Lot NumberK14R033 Certificate TypeCertificate of Analysis Date08/12/2021 Product Code
Lot NumberK16S024 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Alfa Aesar™ 3-(4-Methoxyphenyl)-2-phenylpropionic acid >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.