missing translation for 'onlineSavingsMsg'
Learn More
3,4-Dichlorobenzaldehyde, 98% Product Code.: 10638211

3,4-Dichlorobenzaldehyde, 98%

Product Code. 10638211 Shop All Thermo Scientific Chemicals Products
25 g, Glass bottle
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10638211

Brand: Thermo Scientific Acros 294560250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6287-38-3 | C7H4Cl2O | 175.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6287-38-3
CAS Min % 97.5
CAS Max % 100.0
Melting Point 40.0°C to 44.0°C
Color White to Yellow
Boiling Point 247.0°C to 248.0°C
Flash Point 135°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H4Cl2O
Linear Formula Cl2C6H3CHO
MDL Number MFCD00003351
Quantity 25 g
Beilstein 07, 238
Synonym benzaldehyde, 3,4-dichloro, unii-i6z632wsj5, ccris 6015, pubchem8230, 3,4-dichlorobenzaldehye, 3,4-dichlorobenzaldhyde, 3,4 dichlorobenzaldehyde, 3,4-dichlorobenzaidehyde, 4,3-dichlorobenzaldehyde, dsstox_cid_4971
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ether and acetone,slightly soluble in methanol and amyl ether
InChI Key ZWUSBSHBFFPRNE-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1C=O)Cl)Cl
IUPAC Name 3,4-dichlorobenzaldehyde
Molecular Weight (g/mol) 175.01
PubChem CID 22710
Formula Weight 175.01
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material 3, 4-Dichlorobenzaldehyde, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0409132 Certificate TypeCertificate of Analysis Date19/12/2023 Catalog Number294560250
compliance-icons
Product Identifier
  • 3,4-Dichlorobenzaldehyde
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,4-Dichlorobenzaldehyde, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.