missing translation for 'onlineSavingsMsg'
Learn More
3,4-Dichlorobenzaldehyde, 97% Product Code.: 11309869

3,4-Dichlorobenzaldehyde, 97%

Product Code. 11309869
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11309869

Brand: Thermo Scientific Alfa Aesar B24706.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6287-38-3 | C7H4Cl2O | 175.008 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6287-38-3
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.008
MDL Number MFCD00003351
InChI Key ZWUSBSHBFFPRNE-UHFFFAOYSA-N
Synonym benzaldehyde, 3,4-dichloro, unii-i6z632wsj5, ccris 6015, pubchem8230, 3,4-dichlorobenzaldehye, 3,4-dichlorobenzaldhyde, 3,4 dichlorobenzaldehyde, 3,4-dichlorobenzaidehyde, 4,3-dichlorobenzaldehyde, dsstox_cid_4971
PubChem CID 22710
IUPAC Name 3,4-dichlorobenzaldehyde
SMILES C1=CC(=C(C=C1C=O)Cl)Cl

Specifications

Melting Point 41°C to 44°C
Boiling Point 247°C to 248°C
Flash Point 135°C (275°F)
Quantity 25 g
UN Number UN1759
Beilstein 636379
Sensitivity Air Sensitive
Formula Weight 175.01
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorobenzaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,4-Dichlorobenzaldehyde, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.