missing translation for 'onlineSavingsMsg'
Learn More
3-(3-Hydroxyphenyl)propionic acid, 98+%, Thermo Scientific Chemicals Product Code.: 11304534

3-(3-Hydroxyphenyl)propionic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11304534
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11304534

Brand: Thermo Scientific Alfa Aesar L01279.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-Hydroxyphenylpropionic acid acts as a urinary metabolite of procyanidins in pigs. It also serves as an intermediate in the preparation of various synthetic organic products. Further, it is used in the preparation of 3-(3-hydroxy-phenyl)-propionic acid methyl ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxyphenylpropionic acid acts as a urinary metabolite of procyanidins in pigs. It also serves as an intermediate in the preparation of various synthetic organic products. Further, it is used in the preparation of 3-(3-hydroxy-phenyl)-propionic acid methyl ester.

Solubility
Soluble in chloroform and methanol.

Notes
Incompatible with bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-54-5
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00016554
InChI Key QVWAEZJXDYOKEH-UHFFFAOYSA-N
Synonym 3-3-hydroxyphenyl propanoic acid, 3-3-hydroxyphenyl propionic acid, dihydro-3-coumaric acid, benzenepropanoic acid, 3-hydroxy, 3-m-hydroxyphenyl propionic acid, 3-hydroxybenzenepropanoic acid, 3-hydroxyphenylpropionic acid, m-hydroxyphenylpropionic acid, beta-m-hydroxyphenyl propionic acid, hydrocinnamic acid, m-hydroxy
PubChem CID 91
ChEBI CHEBI:1427
IUPAC Name 3-(3-hydroxyphenyl)propanoic acid
SMILES C1=CC(=CC(=C1)O)CCC(=O)O

Specifications

Melting Point 110°C to 114°C
Quantity 25 g
Beilstein 1947445
Solubility Information Soluble in chloroform and methanol.
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material 3-(3-Hydroxyphenyl)propionic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253695 Certificate TypeCertificate of Analysis Date16/01/2025 Catalog NumberL01279.06,L01279.14
Lot Number10172665 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL01279.06,L01279.14
Lot Number10224963 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL01279.06,L01279.14
Lot Number10136152 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL01279.06,L01279.14
Lot Number10197885 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL01279.06,L01279.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(3-Hydroxyphenyl)propionic acid, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.