missing translation for 'onlineSavingsMsg'
Learn More

3,3-Difluoropiperidine hydrochloride, 99%, Thermo Scientific™

Product Code. 11329735
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496807-97-7
Molecular Formula C5H10ClF2N
Molecular Weight (g/mol) 157.589
MDL Number MFCD03452800
InChI Key LEHHIPIDKQVNEV-UHFFFAOYSA-N
Synonym 3,3-difluoropiperidine hydrochloride, 3,3-difluoropiperidine hcl, piperidine, 3,3-difluoro-, hydrochloride, 3,3-difluoropiperidine, chloride, pubchem10314, acmc-1an4u, 3,3-difluoropiperidine-hcl, 3,3-difluoro-piperidine hcl, 3,3-difluoro-piperidine hydrochloride, 3,3-bis fluoranyl piperidine hydrochloride
PubChem CID 2758349
IUPAC Name 3,3-difluoropiperidine;hydrochloride
SMILES C1CC(CNC1)(F)F.Cl

Specifications

Melting Point ∼240°C
Quantity 1 g
Sensitivity Hygroscopic
Formula Weight 157.59
Percent Purity 99%
Chemical Name or Material 3,3-Difluoropiperidine hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,3-Difluoropiperidine hydrochloride, 99%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.