Learn More
DWK Life Sciences Kimble Chase™ 3.3 Borosilicate Glass Test Tubes
Use round-bottom 3.3 Borosilicate Glass Test Tubes for laboratory needs when heat resistance is critical. These disposable test tubes are thick-walled and autoclavable.
Brand: DWK Life Sciences A620030001221
326.65 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
This item is not returnable.
View return policy
Description
- Disposable
- Borosilicate glass
- Excellent resistance to temperature changes
- Autoclavable
Specifications
Round bottom | |
50Pack | |
Test Tube |
200 mm | |
30 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
31Hours
11Minutes
22Seconds
1
Product Content Correction
Product Title
DWK Life Sciences Kimble Chase™ 3.3 Borosilicate Glass Test Tubes > Diameter: 30mm; Length: 200mm; Quantity: 50/pk