Learn More
Lenz Laborglasinstrumente™ 3.3 Borosilicate Glass Stopcocks
Product Code. 10160581
2.5 mm, Plug size: NS 12.5, Solid Glass Plug, Pack of 10
Click to view available options
Plug Bore:
2.5 mm
4 mm
Description:
Plug size: NS 12.5
Plug size: NS 14.5
Plug size: NS 18.8
Includes:
PTFE Plug, Screw Thread Retaining Device
Solid Glass Plug
Solid Glass Plug, Capillary Side Arms
Unit Size:
Each
Pack of 10
Product Code. 10160581
Brand: Lenz Laborglasinstrumente™ 02101202
Only null left
Specifications
Type Stopcock
Plug Bore 2.5 mm
Description Plug size: NS 12.5
Material Borosilicate Glass
Includes Solid Glass Plug
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ 3.3 Borosilicate Glass Stopcocks >