Learn More
3,3,3-Trifluoro-2-(trifluoromethyl)propionic acid, 97%, Thermo Scientific Chemicals
510.78 NOK - 1475.70 NOK
Chemical Identifiers
CAS | 564-10-3 |
---|---|
Molecular Formula | C4H2F6O2 |
Molecular Weight (g/mol) | 196.05 |
MDL Number | MFCD00464165 |
InChI Key | RAEAYTICAPHWJW-UHFFFAOYSA-N |
Synonym | 3,3,3-trifluoro-2-trifluoromethyl propanoic acid, 2-trifluoromethyl-3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-2-trifluoromethyl propionic acid, propanoic acid,3,3,3-trifluoro-2-trifluoromethyl, hexafluoroisobutyric acid, acmc-1amsz, cf3 2chco2h, 3,3,3-trifluoro-2-trifluoromethylpropionic acid, 3,3,3-trifluoro-2-trifluoromethyl-propionic acid, 3,3,3-tris fluoranyl-2-trifluoromethyl propanoic acid |
PubChem CID | 385606 |
IUPAC Name | 3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid |
SMILES | C(C(=O)O)(C(F)(F)F)C(F)(F)F |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
564-10-3 | |
196.05 | |
RAEAYTICAPHWJW-UHFFFAOYSA-N | |
385606 | |
C(C(=O)O)(C(F)(F)F)C(F)(F)F |
C4H2F6O2 | |
MFCD00464165 | |
3,3,3-trifluoro-2-trifluoromethyl propanoic acid, 2-trifluoromethyl-3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-2-trifluoromethyl propionic acid, propanoic acid,3,3,3-trifluoro-2-trifluoromethyl, hexafluoroisobutyric acid, acmc-1amsz, cf3 2chco2h, 3,3,3-trifluoro-2-trifluoromethylpropionic acid, 3,3,3-trifluoro-2-trifluoromethyl-propionic acid, 3,3,3-tris fluoranyl-2-trifluoromethyl propanoic acid | |
3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid |
Specifications
564-10-3 | |
100.0 | |
White | |
97% | |
(CF3)2CHCOOH | |
3,3,3-trifluoro-2-trifluoromethyl propanoic acid, 2-trifluoromethyl-3,3,3-trifluoropropionic acid, 3,3,3-trifluoro-2-trifluoromethyl propionic acid, propanoic acid,3,3,3-trifluoro-2-trifluoromethyl, hexafluoroisobutyric acid, acmc-1amsz, cf3 2chco2h, 3,3,3-trifluoro-2-trifluoromethylpropionic acid, 3,3,3-trifluoro-2-trifluoromethyl-propionic acid, 3,3,3-tris fluoranyl-2-trifluoromethyl propanoic acid | |
C(C(=O)O)(C(F)(F)F)C(F)(F)F | |
196.05 | |
196.05 | |
Crystalline Powder |
96.0 | |
50°C to 53°C | |
Authentic | |
C4H2F6O2 | |
MFCD00464165 | |
RAEAYTICAPHWJW-UHFFFAOYSA-N | |
3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid | |
385606 | |
97% | |
3, 3, 3-Trifluoro-2-(trifluoromethyl)propionic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and
GHS Signal Word: Warning
- 3,3,3-Trifluoro-2-(trifluoromethyl)propionic acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.