missing translation for 'onlineSavingsMsg'
Learn More
3-(2-Methoxy-3-pyridyl)acrylic acid, 95% Product Code.: 11484460

3-(2-Methoxy-3-pyridyl)acrylic acid, 95%

Product Code. 11484460
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11484460

Brand: Thermo Scientific Alfa Aesar H50063.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 131674-41-4 | C9H9NO3 | 179.175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131674-41-4
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD07781156
InChI Key MTVRDAQJMCORAQ-SNAWJCMRSA-N
Synonym 3-2-methoxy-3-pyridyl acrylic acid, e-3-2-methoxypyridin-3-yl acrylic acid, 3-2-methoxypyridin-3-yl acrylic acid, 2e-3-2-methoxypyridin-3-yl prop-2-enoic acid, 2e-3-2-methoxy 3-pyridyl prop-2-enoic acidShow MoreShow Less
PubChem CID 14691926
IUPAC Name (E)-3-(2-methoxypyridin-3-yl)prop-2-enoic acid
SMILES COC1=C(C=CC=N1)C=CC(=O)O

Specifications

Quantity 5 g
Formula Weight 179.18
Percent Purity 95%
Chemical Name or Material 3-(2-Methoxy-3-pyridyl)acrylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberD21W008 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50063.03,H50063.06
Lot NumberI16Q20 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH50063.03,H50063.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(2-Methoxy-3-pyridyl)acrylic acid, 95% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.