Learn More
3M™ 2890 Series Polycarbonate Lens Safety Goggles
Brand:  3M™ 7000032480
1350.07 NOK valid until 2025-06-08
Use promo code "24131" to get your promotional price.
 View more versions of this product
Product Code. 13458997
  • 2870.00 NOK / Pack of 10
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Specifications
EN 166:2001
Clear
Construction
Indirect ventilation
Yes
Scratch-resistant
Safety goggles
1 Ea.
Polycarbonate
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
    New Offer
    Shop Now
Product Content Correction
Product Title
3M™ 2890 Series Polycarbonate Lens Safety Goggles > Lens Tint: Clear; Lens Coating: Anti-scratch