Learn More
Fisher Chemical Sodium THIOSULFATE - CERTIFIED 25kg
Brand: Fisher Chemical S/7240/70
12814.17 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Sodium THIOSULFATE - CERTIFIED 25kg
This item is not returnable.
View return policy
Product Title
Fisher Chemical Sodium THIOSULFATE - CERTIFIED 25kg >