Learn More
Macherey-Nagel™ 25 Position Container
Brand: Macherey-Nagel™ 702516
This item is not returnable.
View return policy
Specifications
Blue | |
Closure | |
Lid | |
80 mm |
Product Content Correction
Product Title
Macherey-Nagel™ 25 Position Container > Color: Blue