Learn More
uvex 2112 Xtra-Fit Disposable Ear Plugs
Ergonomic, pre-formed ear plugs expand slowly to adapt to the size and shape of the ear canal, thus minimizing leakage and ensuring optimal protection.
Brand: uvex 2112061
477.92 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Ergonomically, pre-shaped disposable earplugs
- Slowly expand to adapt to the size and shape of the ear canal
- Minimized leakage for optimal protection
- Suitable for larger ear canals
- Available in hygienic polybags, or refill bags
Specifications
Orange | |
Large | |
Refill Box | |
36 dB | |
Oval | |
Single-use | |
No |
No | |
2112 Xtra-Fit | |
250 | |
Uncorded | |
Ear Plugs | |
No | |
EN 352-2 |