Learn More
uvex 2112 One-Fit Disposable Ear Plugs
Characterized by their ergonomic, closed surface design One-Fit ear plugs are constructed of polyurethane foam for optimal comfort, even during long periods of use.
Brand: uvex 2112046
317.99 NOK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Soft, polyurethane foam ensures comfort even during long periods of use
- Closed surface prevents foreign bodies from entering
- For use with noise sources up to 110 db(A)
- Cordless, ergonomic design
Specifications
Turquoise | |
Regular | |
Refill Bag | |
37 dB | |
Oval | |
Single-use | |
No |
No | |
2112 One-Fit | |
200 | |
Uncorded | |
Ear Plugs | |
No | |
EN 352-2 |