Learn More
Fisher Chemical 200LT 2.2.4-TRIMETHYLPENTANE - SPECIFIED
Brand: Fisher Chemical T/3600/27
34148.74 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
200LT 2.2.4-TRIMETHYLPENTANE - SPECIFIED
Description
Safety and Handling
- 2,2,4-Trimethylpentane
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P331-Do NOT induce vomiting.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2541918 | Certificate of Analysis | 20/06/2025 | T/3600/08,T/3600/21,T/3600/27,T/3600/17,T/3600/25,T/3600/PB17 |
2535091 | Certificate of Analysis | 24/05/2025 | T/3600/08,T/3600/21,T/3600/27,T/3600/17,T/3600/25,T/3600/PB17 |
2534592 | Certificate of Analysis | 24/05/2025 | T/3600/08,T/3600/21,T/3600/27,T/3600/17,T/3600/25,T/3600/PB17 |
2530560 | Certificate of Analysis | 24/05/2025 | T/3600/08,T/3600/21,T/3600/27,T/3600/17,T/3600/25,T/3600/PB17 |
2528581 | Certificate of Analysis | 24/05/2025 | T/3600/08,T/3600/21,T/3600/27,T/3600/17,T/3600/25,T/3600/PB17 |