Learn More
RETSCH 200 dia. x 25mmH Stainless Steel Test Sieve
Brand:  RETSCH 60.123.001000
 View more versions of this product
Product Code. 10660272
  • 1390.00 NOK / Each
Only null left
Estimated Shipment: 19-09-2025
Specifications
1Each
Test Sieve
Stainless Steel
25 mm
200 mm
Stainless Steel
1 mm
ASTM E 11
Shaker AS 300
For separation, fractioning, particle size determination
Stainless Steel
Certificates
Promotions
Promotions
Product Content Correction
Product Title
RETSCH 200 dia. x 25mmH Stainless Steel Test Sieve > Pore Size: 1 mm