Learn More
Ohaus™ 2 x Centrifuge Racks, Frontier™ R-S4x100/5MIS Swing Out Rotors
Fit inside a swing-out bucket to facilitate centrifugation of tubes of custom size/number.
Brand:  Ohaus™ 30314865
2006.34 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15816702
  • 2065.00 NOK / Pack of 2
Explore more special offers
Description
Description
  • Allows you to centrifuge tubes of custom size or number
  • Suitable for use in buckets for Frontier™ R-S4x100/5MIS Swing Out Rotors
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 x 85 mL
R-S4x100/5MIS Swing Out Rotors
1
38
Rack, 1x85ml D38mm RB, 2/pk, V2
2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Ohaus™ 2 x Centrifuge Racks, Frontier™ R-S4x100/5MIS Swing Out Rotors > 1 x 85 mL