Learn More
2-(Tri-n-butylstannyl)furan, 97% Product Code.: 15451448
2-(Tri-n-butylstannyl)furan, 97%
Product Code. p-7052674
5 mL, 5mL
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Unit Size:
100mL
25mL
5mL
Product Code. 15451448
Brand: Thermo Scientific Alfa Aesar H55259.AA
Request Bulk or Custom Format
CAS: 118486-94-5 | C16H30OSn | 357.125 g/mol
It is an agrochemical, organic and pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an agrochemical, organic and pharmaceutical intermediates.
Notes
Store at room temperature. Light sensitive.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 118486-94-5
Molecular Formula C16H30OSn
Molecular Weight (g/mol) 357.125
MDL Number MFCD00192512
InChI Key SANWDQJIWZEKOD-UHFFFAOYSA-N
Synonym 2-tributylstannyl furan, tributyl furan-2-yl stannane, 2-tributylstannylfuran, tributyl 2-furyl tin, tributyl 2-furyl stannane, 2-tri-n-butylstannyl furan, stannane,tributyl-2-furanyl, stannane, tributyl-2-furanyl, 2-furyltributyltin, tributylstannylfuran
PubChem CID 4393688
IUPAC Name tributyl(furan-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CO1
Specifications
Color Colorless to Yellow
Density 1.134 g/mL
Boiling Point 90°C (0.1 mmHg)
Refractive Index 1.494
Quantity 5 mL
UN Number UN2788
Formula Weight 357.1
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-(Tri-n-butylstannyl)furan
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberY03I021 Certificate TypeCertificate of Analysis Date09/11/2022 Product CodeH55259.AE,H55259.AC,H55259.AA
Lot NumberX04I056 Certificate TypeCertificate of Analysis Date13/10/2022 Product CodeH55259.AE,H55259.AC,H55259.AA
Lot NumberP09H019 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH55259.AE,H55259.AC,H55259.AA
Lot Number5005K07V Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH55259.AE,H55259.AC,H55259.AA
Lot NumberP23H041 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH55259.AE,H55259.AC,H55259.AA
Product Title
2-(Tri-n-butylstannyl)furan, 97% >