missing translation for 'onlineSavingsMsg'
Learn More
2-Propanol, Synthesis Grade, Scharlab Product Code.: 11320456
compliance-icons compliance-icons

2-Propanol, Synthesis Grade, Scharlab

Product Code. 11320456
5 L
Click to view available options
Quantity:
5 L
This item is not returnable. View return policy

Product Code. 11320456

Brand: Scharlab AL0310005P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable for use in solvents for liquid chromatography, in antifreeze compositions and in cosmetics.

  • Synonyms: Isopropyl alcohol, Isopropanol, iso-Propanol, Dimethylcarbinol, 2-Hydroxypropane
  • Molecular Formula: C3H8O
  • Molar Mass: 60.10g/mol
  • CAS: 67-63-0
  • EIN ECS No.: 200-661-7
  • Dielectric Constant: 18.3 at 25°C

Specifications

Melting Point -89.5°C
CAS 67-63-0
Color Colorless
Density 0.78g/cm3
Boiling Point 82.4°C
Quantity 5 L
Flash Point 12°C
Identification Passes Test
Ignition Point 425°C
Physical Form Liquid
Residue after Evaporation 0.005% max
Molecular Formula C3H8O
Vapor Pressure 43hPa at 20°C
Water 0.1% max
Synonym isopropanol, 2-propanol, isopropyl alcohol, 2-hydroxypropane, alkolave, avantine, hartosol, dimethylcarbinol, sec-propyl alcohol, petrohol
Solubility Information Miscible in water (20°C)
InChI Key KFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILES CC(C)O
IUPAC Name propan-2-ol
Molecular Weight (g/mol) 60.096
PubChem CID 3776
ChEBI CHEBI:17824
Formula Weight 60.1g/mol
Percent Purity 99.5% min
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Propanol, synthesis grade
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Propanol, Synthesis Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.