Learn More
Lenz Laborglasintrumente 2-Neck Adapters
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
325.00 NOK - 466.00 NOK
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | ||||||
10221411
|
Lenz Laborglasintrumente
05000111 |
Socket Vertical NS 14/23 |
325.00 NOK
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
10244581
|
Lenz Laborglasintrumente
05000222 |
Socket Vertical NS 19/26 |
466.00 NOK
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
10366071
|
Lenz Laborglasintrumente
05000414 |
Socket Vertical NS 29/32 |
385.00 NOK
Each |
Estimated Shipment: 08-05-2025
Log in to see stock. |
||||||
Description
- With two NS sockets and one NS cone
- Ascending angle: 105°
- Made of DURAN™ tubing
Specifications
Duran Glass |
Product Title
Lenz Laborglasintrumente 2-Neck Adapters