Learn More
2-(Methylthio)ethanol, 99%, Thermo Scientific Chemicals Product Code.: 11387445
2-(Methylthio)ethanol, 99%, Thermo Scientific Chemicals
Product Code. 11387445
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
Product Code. 11387445
Brand: Thermo Scientific Alfa Aesar L05442.14
Request Bulk or Custom Format
2-(Methylthio)ethanol is used as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(Methylthio)ethanol is used as an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Not miscible in water.
Notes
Do not store together with acids. Store away from Oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5271-38-5
Molecular Formula C3H8OS
Molecular Weight (g/mol) 92.16
MDL Number MFCD00002908
InChI Key WBBPRCNXBQTYLF-UHFFFAOYSA-N
Synonym 2-methylthio ethanol, methylthioethanol, ethanol, 2-methylthio, 2-hydroxyethyl methyl sulfide, 2-methylmercaptoethanol, 2-methylsulfanyl ethanol, 2-methylsulfanyl-ethanol, beta-hydroxyethyl methyl sulfide, 2-methylthioethanolShow MoreShow Less
PubChem CID 78925
ChEBI CHEBI:63861
IUPAC Name 2-methylsulfanylethanol
SMILES CSCCO
Specifications
Density 1.06
Boiling Point 169°C to 171°C
Flash Point 70°C (158°F)
Odor Stench
Linear Formula CH3S(CH2)2OH
Refractive Index 1.4944
Quantity 25 g
UN Number UN3334
Beilstein 1731081
Solubility Information Not miscible in water.
Formula Weight 92.16
Percent Purity 99%
Chemical Name or Material 2-(Methylthio)ethanol
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250908 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL05442.06,L05442.14,L05442.22
Lot Number10218004 Certificate TypeCertificate of Analysis Date22/04/2024 Catalog NumberL05442.06,L05442.14,L05442.22
Lot Number10242959 Certificate TypeCertificate of Analysis Date23/08/2022 Catalog NumberL05442.06,L05442.14,L05442.22
Lot Number10192991 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL05442.06,L05442.14,L05442.22
Lot Number10218004 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL05442.06,L05442.14,L05442.22
Product Title
2-(Methylthio)ethanol, 99%, Thermo Scientific Chemicals >