missing translation for 'onlineSavingsMsg'
Learn More
2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride, 97%, Thermo Scientific Chemicals Product Code.: 15458955

2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 15458955
1 g, 1g
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15458955

Brand: Thermo Scientific Alfa Aesar H34468.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use as chiral derivatizing agents in the determination of the enantiomeric excess of alcohols and amines. They are widely used as important intermediates in the synthesis of dyes, drugs and pigments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Use as chiral derivatizing agents in the determination of the enantiomeric excess of alcohols and amines. They are widely used as important intermediates in the synthesis of dyes, drugs and pigments.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture. Water, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1257535-05-9
Molecular Formula C7H5ClF3NO2S
Molecular Weight (g/mol) 259.63
MDL Number MFCD14583150
InChI Key BNCNLCKWRCXBQH-UHFFFAOYSA-N
Synonym 2-methyl-6-trifluoromethyl pyridine-3-sulfonyl chloride, 3-chlorosulphonyl-2-methyl-6-trifluoromethyl pyridine
PubChem CID 51342031
IUPAC Name 2-methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride
SMILES CC1=C(C=CC(=N1)C(F)(F)F)S(Cl)(=O)=O

Specifications

Melting Point 57°C to 59°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 259.64
Percent Purity 97%
Chemical Name or Material 2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Product Code
Lot Number10174157 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH34468.MD,H34468.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Methyl-6-(trifluoromethyl)pyridine-3-sulfonyl chloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.