Learn More
2-Methyl-1-phenyl-2-propanol, 98+%, Thermo Scientific Chemicals
598.00 NOK - 2040.00 NOK
Chemical Identifiers
CAS | 100-86-7 |
---|---|
Molecular Formula | C10H14O |
Molecular Weight (g/mol) | 150.221 |
MDL Number | MFCD00004465 |
InChI Key | RIWRBSMFKVOJMN-UHFFFAOYSA-N |
Synonym | 2-methyl-1-phenyl-2-propanol, benzyldimethylcarbinol, 2-benzyl-2-propanol, phenyl-tert-butanol, dimethylbenzylcarbinol, dmbc, 1,1-dimethylphenylethanol, 1,1-dimethyl-2-phenylethanol, benzylpropyl alcohol, dimethyl benzyl carbinolShow More |
PubChem CID | 7531 |
IUPAC Name | 2-methyl-1-phenylpropan-2-ol |
SMILES | CC(C)(CC1=CC=CC=C1)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11382256
|
Thermo Scientific Alfa Aesar
L03679.22 |
100 g |
598.00 NOK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11392256
|
Thermo Scientific Alfa Aesar
L03679.36 |
500 g |
2040.00 NOK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Methyl-1-phenyl-2-propanol is used in confecting floral essences, such as lily, narcissus, jasmine and keiskei and other advanced floral essences. Its acetic ester has fresh fragrance so that it has especial value.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Methyl-1-phenyl-2-propanol is used in confecting floral essences, such as lily, narcissus, jasmine and keiskei and other advanced floral essences. Its acetic ester has fresh fragrance so that it has especial value.
Solubility
Slightly soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
100-86-7 | |
150.221 | |
RIWRBSMFKVOJMN-UHFFFAOYSA-N | |
7531 | |
CC(C)(CC1=CC=CC=C1)O |
C10H14O | |
MFCD00004465 | |
2-methyl-1-phenyl-2-propanol, benzyldimethylcarbinol, 2-benzyl-2-propanol, phenyl-tert-butanol, dimethylbenzylcarbinol, dmbc, 1,1-dimethylphenylethanol, 1,1-dimethyl-2-phenylethanol, benzylpropyl alcohol, dimethyl benzyl carbinolShow More | |
2-methyl-1-phenylpropan-2-ol |
Specifications
100-86-7 | |
0.974 | |
82°C (179°F) | |
1.515 | |
1855608 | |
Slightly soluble in water. | |
CC(C)(CC1=CC=CC=C1)O | |
150.221 | |
150.22 | |
2-Methyl-1-phenyl-2-propanol |
23°C to 25°C | |
215°C | |
C10H14O | |
MFCD00004465 | |
2-methyl-1-phenyl-2-propanol, benzyldimethylcarbinol, 2-benzyl-2-propanol, phenyl-tert-butanol, dimethylbenzylcarbinol, dmbc, 1,1-dimethylphenylethanol, 1,1-dimethyl-2-phenylethanol, benzylpropyl alcohol, dimethyl benzyl carbinol | |
RIWRBSMFKVOJMN-UHFFFAOYSA-N | |
2-methyl-1-phenylpropan-2-ol | |
7531 | |
≥98% |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 202-896-0
missing translation for 'rtecsNumber' : SG8050000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.