missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethanol, ACS Reagent, ≥99.3%, Honeywell™

Product Code. 15685070
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 109-86-4
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
MDL Number MFCD00002867
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
PubChem CID 8019
ChEBI CHEBI:46790
IUPAC Name 2-methoxyethanol
SMILES COCCO

Specifications

Melting Point -85°C
Density 0.965g/mL at 25°C
Boiling Point 125°C
Explosion Limit 0.245
Flash Point 40°C (104°F)
Ignition Point 548°F
Infrared Spectrum Conforms to structure
Odor Characteristic
Packaging Glass Bottle
Linear Formula CH3OCH2CH2OH
Quantity 1 L
UN Number UN1188
Beilstein 1731074
Stabilizer 50ppm BHT
Formula Weight 76.09g/mol
Vapor Pressure 6.17mmHg (20°C)
Percent Purity ≥99.3%
Physical Form Liquid
Chemical Name or Material 2-Methoxyethanol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.