missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-(trifluoromethoxy)benzaldehyde, 98%, Thermo Scientific™

Product Code. 15497545
250 mg, 250mg
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
missing translation for 'unitSize'
1g
250mg
This item is not returnable. View return policy

Product Code. 15497545

missing translation for 'mfr': Thermo Scientific Alfa Aesar H26105.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145742-65-0
Molecular Formula C9H7F3O3
Molecular Weight (g/mol) 220.15
MDL Number MFCD04115958
InChI Key ATRDCTRZAJKDPL-UHFFFAOYSA-N
Synonym 2-methoxy-5-trifluoromethoxy benzaldehyde, 2-formyl-4-trifluoromethoxy anisole, 2-methoxy-5-trifluoromethoxy-benzaldehyde, acmc-209cvk, 2-methoxy-5-trifluoromethoxybenzaidehyde, 2-methoxy-5-trifluoromethyloxy benzaldehyde, benzaldehyde,2-methoxy-5-trifluoromethoxy, benzaldehyde, 2-methoxy-5-trifluoromethoxyShow MoreShow Less
PubChem CID 2783364
IUPAC Name 2-methoxy-5-(trifluoromethoxy)benzaldehyde
SMILES COC1=CC=C(OC(F)(F)F)C=C1C=O

Specifications

Melting Point 20°C to 25°C
Density 1.36
Refractive Index 1.475
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 220.15
Percent Purity 98%
Chemical Name or Material 2-Methoxy-5-(trifluoromethoxy)benzaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Product Code
Lot Number10146244 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Lot Number10146245 Certificate TypeCertificate of Analysis Date06/12/2021 Product Code
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Methoxy-5-(trifluoromethoxy)benzaldehyde, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.