missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Mercaptoethanol, ≥98%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1 L
Unit Size:
1 L
100mL
250mL
500mL
Specifications
Specifications
Color | Colorless to Yellow |
Density | 1.12g/cm3 at 20°C |
Assay Percent Range | ≥98% |
Quantity | 500 mL |
IUPAC Name | 2-sulfanylethan-1-ol |
Formula Weight | 78.13 |
Physical Form | Liquid |
Chemical Name or Material | 2-Mercaptoethanol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2-Mercaptoethanol, ≥98%, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction