missing translation for 'onlineSavingsMsg'
Learn More
2-Hydrazino-2-imidazoline hydrobromide, 98%, Thermo Scientific Chemicals Product Code.: 11404307

2-Hydrazino-2-imidazoline hydrobromide, 98%, Thermo Scientific Chemicals

Product Code. 11404307
10 g
Click to view available options
Quantity:
2.5 g
10 g
This item is not returnable. View return policy

Product Code. 11404307

Brand: Thermo Scientific Alfa Aesar B20386.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55959-84-7
Molecular Formula C3H9BrN4
Molecular Weight (g/mol) 181.037
MDL Number MFCD00012694
InChI Key HIJRJVVSABWXKG-UHFFFAOYSA-N
Synonym 2-hydrazino-2-imidazoline hydrobromide, 2-hydrazinyl-4,5-dihydro-1h-imidazole hydrobromide, 2-hydrazino-delta-2-imidazoline hydrobromide, 4,5-dihydroimidazol-2-ylhydrazine hydrobromide, 4,5-dihydro-2-hydrazino-1h-imidazole hydrobromide, 2-hydrazino-4,5-dihydro-1h-imidazole hydrobromide, 4,5-dihydro-1h-imidazol-2-ylhydrazine hydrobromide, 4,5-dihydro-1h-imidazol-2-yl-hydrazine hydrobromide, 4,5-dihydro-1h-imidazol-2-yl-hydrazine, acmc-1av1kShow MoreShow Less
PubChem CID 3085099
IUPAC Name 4,5-dihydro-1H-imidazol-2-ylhydrazine;hydrobromide
SMILES C1CN=C(N1)NN.Br

Specifications

Melting Point 181°C to 185°C
Quantity 10 g
Sensitivity Hygroscopic
Formula Weight 181.04
Percent Purity 98%
Chemical Name or Material 2-Hydrazino-2-imidazoline hydrobromide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10191607 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20386.09,B20386.88
Lot Number10131175 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20386.09,B20386.88
Lot Number10198810 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20386.09,B20386.88
Lot Number10226682 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20386.09,B20386.88
Lot Number10211405 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20386.09,B20386.88

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Hydrazino-2-imidazoline hydrobromide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.