Learn More
Fisherbrand™ 2 Hole Natural Rubber Stopper
Prevents liquid chemical leaks or escape from container
299.00 NOK - 451.00 NOK
Specifications
Color | Red |
---|---|
Material | Natural Rubber |
Product Type | Stopper (2 holes) |
Hardness | 46/55 IRHD |
Height (Metric) | 54mm |
Product Code | Brand | Diameter (Metric) Bottom | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Bottom | Price | Quantity & Availability | ||||||
11577642
|
Fisherbrand™
RB092/2H |
92 mm |
345.00 NOK
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||||
11567642
|
Fisherbrand™
RB079/2H |
79 mm |
299.00 NOK
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||||
11597642
|
Fisherbrand™
RB105/2H |
105 mm |
451.00 NOK
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||||
Specifications
Red | |
Stopper (2 holes) | |
54mm |
Natural Rubber | |
46/55 IRHD | |
1/Ea. |
Product Title
Fisherbrand™ 2 Hole Natural Rubber Stopper