missing translation for 'onlineSavingsMsg'
Learn More
2-Heptyn-1-ol, 97%, Thermo Scientific Chemicals Product Code.: 11407167

2-Heptyn-1-ol, 97%, Thermo Scientific Chemicals

Product Code. 11407167
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11407167

Brand: Thermo Scientific Alfa Aesar A14959.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1002-36-4
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00039541
InChI Key OGDYROFIIXDTQK-UHFFFAOYSA-N
Synonym 2-heptyn-1-ol, 2-heptyne-1-ol, 2-heptynol, pubchem13065, acmc-2097no, hoch2c=c ch2 3ch3
PubChem CID 535111
IUPAC Name hept-2-yn-1-ol
SMILES CCCCC#CCO

Specifications

Density 0.882
Boiling Point 89°C to 90°C (16 mmHg)
Flash Point 78°C (172°F)
Refractive Index 1.454
Quantity 25 g
Beilstein 1739001
Formula Weight 112.17
Percent Purity 97%
Chemical Name or Material 2-Heptyn-1-ol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10219415 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14959.06,A14959.14
Lot Number10214005 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14959.06,A14959.14
Lot Number10206297 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14959.06,A14959.14
Lot Number10044280 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14959.06,A14959.14
Lot Number10181291 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14959.06,A14959.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Heptyn-1-ol, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.