missing translation for 'onlineSavingsMsg'
Learn More
2-Fluoropyridine-4-boronic acid pinacol ester, 95% Product Code.: 11466330

2-Fluoropyridine-4-boronic acid pinacol ester, 95%

Product Code. 11466330
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11466330

Brand: Thermo Scientific Alfa Aesar H27179.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 458532-86-0 | C11H15BFNO2 | 223.05 g/mol

Used in pharmaceuticals, agrochemicals, and high-tech materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in pharmaceuticals, agrochemicals, and high-tech materials.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 458532-86-0
Molecular Formula C11H15BFNO2
Molecular Weight (g/mol) 223.05
MDL Number MFCD06798253
InChI Key PCLMNCBIXQQRMB-UHFFFAOYSA-N
Synonym 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridine-4-boronic acid pinacol ester, 2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridin-4-boronic acid pinacol ester, 2-fluoropyridin-4-ylboronic acid pinacol ester, pyridine, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, abbypharma ap-11-5396, 2-fluoro-4-pyridineboronic acid pinacol ester, 2-fluoropyridin-4-yl boronic acid pinacol ester, 2-2-fluoro 4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 12060164
IUPAC Name 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=CC(F)=NC=C1

Specifications

Melting Point 48°C to 52°C
Quantity 1 g
Formula Weight 223.06
Percent Purity 95%
Chemical Name or Material 2-Fluoropyridine-4-boronic acid pinacol ester

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS07C044 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27179.06,H27179.03
Lot Number10143547 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27179.06,H27179.03
Lot Number10151510 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27179.06,H27179.03
Lot Number10170859 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27179.06,H27179.03
Lot Number10143050 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27179.06,H27179.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Fluoropyridine-4-boronic acid pinacol ester, 95% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.