missing translation for 'onlineSavingsMsg'
Learn More
2-Cyano-4'-methylbiphenyl, 98+%, Thermo Scientific Chemicals Product Code.: 11369629

2-Cyano-4'-methylbiphenyl, 98+%, Thermo Scientific Chemicals

Product Code. 11369629
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11369629

Brand: Thermo Scientific Alfa Aesar B21273.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 114772-53-1 | C14H11N | 193.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114772-53-1
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.25
MDL Number MFCD00151805
InChI Key ZGQVZLSNEBEHFN-UHFFFAOYSA-N
Synonym 2-cyano-4'-methylbiphenyl, 4'-methyl-2-cyanobiphenyl, 4'-methylbiphenyl-2-carbonitrile, 2'-cyano-4-methylbiphenyl, 2-p-tolyl benzonitrile, 2-4-methylphenyl benzonitrile, 2-cyano-4'-methyl biphenyl, 4'-methyl-1,1'-biphenyl-2-carbonitrile, unii-o417ti15ta, 4'-methyl-2-cyano diphenyl
PubChem CID 145512
IUPAC Name 2-(4-methylphenyl)benzonitrile
SMILES CC1=CC=C(C=C1)C1=CC=CC=C1C#N

Specifications

Melting Point 49°C to 53°C
Density 1.17
Boiling Point >320°C
Quantity 100 g
UN Number UN3077
Beilstein 3605954
Formula Weight 193.26
Percent Purity ≥98%
Chemical Name or Material 2-Cyano-4'-methylbiphenyl

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10157712 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21273.22,B21273.14,B21273.06
Lot Number5006V11V Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21273.22,B21273.14,B21273.06
Lot Number10034067 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21273.22,B21273.14,B21273.06
Lot NumberH2836A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21273.22,B21273.14,B21273.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Cyano-4'-methylbiphenyl, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.