missing translation for 'onlineSavingsMsg'
Learn More
2-Chloropyridine-3-carboxaldehyde, 98% Product Code.: 15450168

2-Chloropyridine-3-carboxaldehyde, 98%

Product Code. 15450168
5 g, 5g
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price

Product Code. 15450168

Brand: Thermo Scientific Alfa Aesar H53399.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price

CAS: 36404-88-3 | C6H4ClNO | 141.554 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36404-88-3
Molecular Formula C6H4ClNO
Molecular Weight (g/mol) 141.554
MDL Number MFCD01315308
InChI Key KHPAGGHFIDLUMB-UHFFFAOYSA-N
Synonym 2-chloro-3-formylpyridine, 2-chloronicotinaldehyde, 2-chloro-3-pyridinecarboxaldehyde, 2-chloro-pyridine-3-carbaldehyde, chloro-2-formyl-3-pyridine, 2-chloropyridine-3-carboxaldehyde, 3-pyridinecarboxaldehyde, 2-chloro, 2-chloro-3-pyridinecarbaldehyde, 2-chloro-3-pyridinecarboxyaldehyde, 2-chloro-pyridin-3-carbaldehyde
PubChem CID 737607
IUPAC Name 2-chloropyridine-3-carbaldehyde
SMILES C1=CC(=C(N=C1)Cl)C=O

Specifications

Melting Point 50°C to 54°C
Flash Point >110°C (230°F)
Quantity 5 g
Formula Weight 141.56
Percent Purity 98%
Chemical Name or Material 2-Chloropyridine-3-carboxaldehyde

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberP28E071 Certificate TypeCertificate of Analysis Date10/06/2025 Product Code
Lot NumberY08E042 Certificate TypeCertificate of Analysis Date22/05/2025 Product CodeH53399.14,H53399.03,H53399.06
Lot NumberD30Z042 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH53399.03,H53399.14,H53399.06
Lot NumberM24E037 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH53399.03,H53399.14,H53399.06
Lot NumberY08E042 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH53399.03,H53399.14,H53399.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloropyridine-3-carboxaldehyde, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.