missing translation for 'onlineSavingsMsg'
Learn More
2-Chloro-6-methylpyridine, 98%, Thermo Scientific Chemicals Product Code.: 11419606

2-Chloro-6-methylpyridine, 98%, Thermo Scientific Chemicals

Product Code. 11419606
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11419606

Brand: Thermo Scientific Alfa Aesar B20174.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18368-63-3
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.57
MDL Number MFCD00006245
InChI Key GXZDYRYYNXYPMQ-UHFFFAOYSA-N
Synonym 6-chloro-2-picoline, 6-chloro-2-methylpyridine, pyridine, 2-chloro-6-methyl, 2-chloro-6-picoline, 2-picoline, 6-chloro, unii-39mx1lq68l, 2-chloro-6-methyl pyridine, 2-chloro-6-methyl-pyridine, 2-chloro-6-methy pyridineShow MoreShow Less
PubChem CID 87601
IUPAC Name 2-chloro-6-methylpyridine
SMILES CC1=CC=CC(Cl)=N1

Specifications

Density 1.167
Boiling Point 64°C to 68°C (10 mmHg)
Flash Point 73°C (163°F)
Refractive Index 1.5265
Quantity 5 g
UN Number UN2810
Beilstein 107187
Formula Weight 127.57
Percent Purity 98%
Chemical Name or Material 2-Chloro-6-methylpyridine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10164147 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20174.06,B20174.14
Lot NumberGBFA021501 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20174.06,B20174.14
Lot Number10136699 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20174.06,B20174.14
Lot NumberH5798C Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20174.06,B20174.14
Lot Number10199824 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20174.06,B20174.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloro-6-methylpyridine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.