missing translation for 'onlineSavingsMsg'
Learn More
2-Chloro-5,5-dimethyl-1,3,2-dioxaphosphorinane, 97%, AcroSeal™, Thermo Scientific™ Product Code.: 10736485

2-Chloro-5,5-dimethyl-1,3,2-dioxaphosphorinane, 97%, AcroSeal™, Thermo Scientific™

Product Code. 10736485
50 mL, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy

Product Code. 10736485

Brand: Thermo Scientific Acros 427840500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

 

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2428-06-0
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2150g/mL
Boiling Point 46.0°C to 47.0°C (1.6 mmHg)
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging AcroSeal™ Glass Bottle
Molecular Formula C5H10ClO2P
Refractive Index 1.4740 to 1.4760 (20°C, 589nm)
MDL Number MFCD03093988
Quantity 50 mL
Specific Gravity 1.215
Synonym 2-chloro-5,5-dimethyl-1,3,2-dioxaphosphorinane, 1,3,2-dioxaphosphorinane, 2-chloro-5,5-dimethyl, phosphorochloridous acid, cyclic 2,2-dimethyltrimethylene ester, 2-chloro-5,5-dimethyl 1,3,2 dioxaphosphinane, 5,5-dimethyl-2-chloro-1,3,2-dioxaphosphorinane, 1,3,2-dioxaphosphorinane,2-chloro-5,5-dimethyl
InChI Key RAOZVFGTRXLBTA-UHFFFAOYSA-N
SMILES CC1(C)COP(Cl)OC1
IUPAC Name 2-chloro-5,5-dimethyl-1,3,2-dioxaphosphinane
Molecular Weight (g/mol) 168.56
PubChem CID 75508
Formula Weight 168.56
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-chloro-5,5-dimethyl-1,3,2-dioxaphosphorinane
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-5,5-dimethyl-1,3,2-dioxaphosphorinane
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloro-5,5-dimethyl-1,3,2-dioxaphosphorinane, 97%, AcroSeal™, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.