missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™

Product Code. 15448395
250 mg, 250mg
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 279252-26-5
Molecular Formula C8H5BrClF3
Molecular Weight (g/mol) 273.477
MDL Number MFCD14155638
InChI Key AIZUUFYFLNVTQX-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl benzyl bromide, 1-bromomethyl-2-chloro-4-trifluoromethyl benzene, 2-chloro-4-trifluoromethylbenzylbromide, 2-chloro-4-trifluoromethyl benzylbromide, benzene,1-bromomethyl-2-chloro-4-trifluoromethy
PubChem CID 22058559
IUPAC Name 1-(bromomethyl)-2-chloro-4-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)Cl)CBr

Specifications

Refractive Index 1.511
Quantity 250 mg
UN Number UN3265
Formula Weight 273.48
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-(trifluoromethyl)benzyl bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloro-4-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.