missing translation for 'onlineSavingsMsg'
Learn More
2-Chloro-4-methoxyphenylacetonitrile, 95% Product Code.: 11361939

2-Chloro-4-methoxyphenylacetonitrile, 95%

Product Code. 11361939
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11361939

Brand: Thermo Scientific Alfa Aesar H26021.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 170737-93-6 | C9H8ClNO | 181.62 g/mol

2-Chloro-4-methoxyphenylacetonitrile is used as a raw material and an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-4-methoxyphenylacetonitrile is used as a raw material and an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170737-93-6
Molecular Formula C9H8ClNO
Molecular Weight (g/mol) 181.62
MDL Number MFCD08445710
InChI Key UKYXNWLFQOJYNQ-UHFFFAOYSA-N
Synonym 2-2-chloro-4-methoxyphenyl acetonitrile, 2-chloro-4-methoxyphenyl acetonitrile, 2-chloro-4-methoxyphenylacetonitrile, acmc-20appn, pubchem11235, 2-chloro-4-methoxyphenylacetnitrile, benzeneacetonitrile, 2-chloro-4-methoxy, 2-2-chloranyl-4-methoxy-phenyl ethanenitrile
PubChem CID 22327506
IUPAC Name 2-(2-chloro-4-methoxyphenyl)acetonitrile
SMILES COC1=CC(Cl)=C(CC#N)C=C1

Specifications

Melting Point 41°C to 43°C
Quantity 1 g
UN Number UN3439
Solubility Information Insoluble in water.
Formula Weight 181.62
Percent Purity 95%
Chemical Name or Material 2-Chloro-4-methoxyphenylacetonitrile

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10156800 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26021.03,H26021.06
Lot Number10143454 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26021.03,H26021.06
Lot Number60900055 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH26021.03,H26021.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Chloro-4-methoxyphenylacetonitrile, 95% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.