missing translation for 'onlineSavingsMsg'
Learn More

2-Carboxyphenyl phosphate, 98%

CAS: 6064-83-1 | C7H7O6P | 218.101 g/mol

707.00 NOK - 10068.00 NOK

Chemical Identifiers

CAS 6064-83-1
Molecular Formula C7H7O6P
Molecular Weight (g/mol) 218.101
MDL Number MFCD00042645
InChI Key FFKUDWZICMJVPA-UHFFFAOYSA-N
Synonym fosfosal, 2-phosphonooxy benzoic acid, disdolen, salicyl phosphate, o-carboxyphenyl phosphate, 2-carboxyphenyl phosphate, 2-phosphonoxybenzoic acid, phosphonoxybenzoic acid, o-phosphonooxy benzoic acid, fosfosal inn
PubChem CID 3418
IUPAC Name 2-phosphonooxybenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)OP(=O)(O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15453087
SDS View Documents
Thermo Scientific Alfa Aesar
A14417.03
1 g
707.00 NOK
1g
Please to purchase this item. Need a web account? Register with us today!
11447047
SDS View Documents
Thermo Scientific Alfa Aesar
A14417.06
5 g
3975.00 NOK
5g
Please to purchase this item. Need a web account? Register with us today!
11457047
SDS View Documents
Thermo Scientific Alfa Aesar
A14417.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

2-Carboxyphenyl phosphate is used as a substrate for the estimation of acid phosphatase. It is also used as a analgesic, anti-inflammatory agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Carboxyphenyl phosphate is used as a substrate for the estimation of acid phosphatase. It is also used as a analgesic, anti-inflammatory agent.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'einecsNumber' : 227-993-5

missing translation for 'rtecsNumber' : VO2770000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Carboxyphenyl phosphate, 98%

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.