Learn More
2-Bromoimidazole, 95%
CAS: 16681-56-4 | C3H3BrN2 | 146.98 g/mol
3260.00 NOK - 11825.00 NOK
Chemical Identifiers
CAS | 16681-56-4 |
---|---|
Molecular Formula | C3H3BrN2 |
Molecular Weight (g/mol) | 146.98 |
MDL Number | MFCD02179526 |
InChI Key | AXHRGVJWDJDYPO-UHFFFAOYSA-N |
Synonym | 2-bromoimidazole, 1h-imidazole, 2-bromo, bromoimidazole, sftheabililuh@, pubchem8987, zlchem 1065, acmc-1btox, 2-bromanyl-1h-imidazole, ksc176e6b, 2-bromo-1h-imidazole |
PubChem CID | 2773261 |
IUPAC Name | 2-bromo-1H-imidazole |
SMILES | BrC1=NC=CN1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15499287
|
Thermo Scientific Alfa Aesar
H52553.03 |
1 g |
3260.00 NOK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15409297
|
Thermo Scientific Alfa Aesar
H52553.06 |
5 g |
11825.00 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Bromoimidazole is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromoimidazole is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Chemical Identifiers
16681-56-4 | |
146.98 | |
AXHRGVJWDJDYPO-UHFFFAOYSA-N | |
2773261 | |
BrC1=NC=CN1 |
C3H3BrN2 | |
MFCD02179526 | |
2-bromoimidazole, 1h-imidazole, 2-bromo, bromoimidazole, sftheabililuh@, pubchem8987, zlchem 1065, acmc-1btox, 2-bromanyl-1h-imidazole, ksc176e6b, 2-bromo-1h-imidazole | |
2-bromo-1H-imidazole |
Specifications
16681-56-4 | |
C3H3BrN2 | |
2-bromoimidazole, 1h-imidazole, 2-bromo, bromoimidazole, sftheabililuh@, pubchem8987, zlchem 1065, acmc-1btox, 2-bromanyl-1h-imidazole, ksc176e6b, 2-bromo-1h-imidazole | |
AXHRGVJWDJDYPO-UHFFFAOYSA-N | |
2-bromo-1H-imidazole | |
2773261 | |
95% |
197°C | |
MFCD02179526 | |
Slightly soluble in water. | |
BrC1=NC=CN1 | |
146.98 | |
146.98 | |
2-Bromoimidazole |
Safety and Handling
GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H335
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.