Learn More
2-Bromo-6-pyridinemethanol, 96% Product Code.: 15432558
2-Bromo-6-pyridinemethanol, 96%
Product Code. 15432558
1 g, 1g
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Product Code. 15432558
Brand: Thermo Scientific Alfa Aesar H56123.03
Request Bulk or Custom Format
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
CAS: 33674-96-3 | C6H6BrNO | 188.02 g/mol
It is an active pharmaceutical ingredient and an pharmaceutical adjuvant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an active pharmaceutical ingredient and an pharmaceutical adjuvant.
Solubility
Slightly soluble in water.
Notes
Incompatible with oxidizing agents and bases. Store at room temperature.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 33674-96-3
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
MDL Number MFCD04039884
InChI Key XDDGKNRSCDEWBR-UHFFFAOYSA-N
Synonym 6-bromopyridin-2-yl methanol, 2-bromo-6-pyridinemethanol, 2-bromo-6-hydroxymethyl pyridine, 6-bromo-2-hydroxymethyl pyridine, 2-pyridinemethanol, 6-bromo, 6-bromopyridine-2-methanol, 6-bromo-pyridine-2-yl methanol, 6-bromo-2-pyridinemethanol, 6-bromo-pyridin-2-yl-methanol, 6-bromo-pyridin-2-yl methanolShow MoreShow Less
PubChem CID 3517811
IUPAC Name (6-bromopyridin-2-yl)methanol
SMILES OCC1=CC=CC(Br)=N1
Specifications
Melting Point 34°C to 39°C
Boiling Point 246°C
Flash Point >110°C (230°F)
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 188.02
Percent Purity 96%
Chemical Name or Material 2-Bromo-6-pyridinemethanol
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberBG250501 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH56123.06,H56123.03
Lot NumberZ11B006 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH56123.06,H56123.03
Lot NumberP27E061 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH56123.06,H56123.03
Lot NumberN17D022 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH56123.06,H56123.03
Lot NumberN07E019 Certificate TypeCertificate of Analysis Date23/11/2021 Product CodeH56123.06,H56123.03
Product Title
2-Bromo-6-pyridinemethanol, 96% >