missing translation for 'onlineSavingsMsg'
Learn More
2-Bromo-5-chloroanisole, 98+%, Thermo Scientific Chemicals Product Code.: 11407196

2-Bromo-5-chloroanisole, 98+%, Thermo Scientific Chemicals

Product Code. 11407196
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11407196

Brand: Thermo Scientific Alfa Aesar B25512.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174913-09-8
Molecular Formula C7H6BrClO
Molecular Weight (g/mol) 221.478
MDL Number MFCD03790889
InChI Key CQGYLDZGJLVLMK-UHFFFAOYSA-N
Synonym 2-bromo-5-chloroanisole, benzene, 1-bromo-4-chloro-2-methoxy, pubchem5265, acmc-1c20c, ksc497i0p, 2-bromo-5-chloroanisole, 98+%, 2-methoxy-4-chloro-1-bromobenzene, 1-bromo-4-chloro-2-methoxy-benzeneShow MoreShow Less
PubChem CID 17984845
IUPAC Name 1-bromo-4-chloro-2-methoxybenzene
SMILES COC1=C(C=CC(=C1)Cl)Br

Specifications

Refractive Index 1.5875
Quantity 25 g
Formula Weight 221.48
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-5-chloroanisole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10226375 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25512.14,B25512.06
Lot Number10129530 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25512.14,B25512.06
Lot Number10141657 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25512.14,B25512.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Bromo-5-chloroanisole, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.