Learn More
2-Benzyloxybenzaldehyde, 98% Product Code.: 11383416
2-Benzyloxybenzaldehyde, 98%
Product Code. p-7046033
10 g, 10g
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. 11383416
Brand: Thermo Scientific Alfa Aesar L12866.09
Request Bulk or Custom Format
CAS: 5896-17-3 | C14H12O2 | 212.25 g/mol
2-Benzyloxybenzaldehyde is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Benzyloxybenzaldehyde is used as an organic chemical synthesis intermediate.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5896-17-3
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00016583
InChI Key PBEJTRAJWCNHRS-UHFFFAOYSA-N
Synonym 2-benzyloxy benzaldehyde, 2-benzyloxybenzaldehyde, benzaldehyde, 2-phenylmethoxy, o-benzyloxy benzaldehyde, benzaldehyde, o-benzyloxy, o-benzylsalicylaldehyde, 2-benzyl-oxy benzaldehyde, 2-phenylmethoxy benzaldehyde, benzyloxybenzaldehyde, o-benzyloxybenzaldehydeShow MoreShow Less
PubChem CID 344784
IUPAC Name 2-phenylmethoxybenzaldehyde
SMILES O=CC1=CC=CC=C1OCC1=CC=CC=C1
Specifications
Melting Point 46°C to 47°C
Quantity 10 g
Beilstein 780307
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 212.25
Percent Purity 98%
Chemical Name or Material 2-Benzyloxybenzaldehyde
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number10251243 Certificate TypeCertificate of Analysis Date30/08/2024 Product CodeL12866.18,L12866.09
Lot Number10248525 Certificate TypeCertificate of Analysis Date02/11/2023 Product CodeL12866.18,L12866.09
Lot Number10113380 Certificate TypeCertificate of Analysis Date06/12/2021 Product CodeL12866.18,L12866.09
Lot Number10167855 Certificate TypeCertificate of Analysis Date06/12/2021 Product CodeL12866.18,L12866.09
Lot Number10174942 Certificate TypeCertificate of Analysis Date02/12/2021 Product CodeL12866.18,L12866.09
Product Title
2-Benzyloxybenzaldehyde, 98% >