missing translation for 'onlineSavingsMsg'
Learn More
2-Aminothiazole, 97% Product Code.: 11409892

2-Aminothiazole, 97%

Product Code. 11409892
100 g, 100g
Click to view available options
Quantity:
50 g
100 g
250 g
500 g
2500 g
Unit Size:
100g
2500g
250g
500g
50g
This item is not returnable. View return policy

Product Code. 11409892

Brand: Thermo Scientific Alfa Aesar A12026.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96-50-4 | C3H4N2S | 100.14 g/mol

2-Aminothiazole is used in the synthesis of 2-aminothiazole-modified silica gel and also used in Ulmann coupling with 2-chlorobenzoic acids mediated by ultrasonic irradiation. It acts as a component of disperse dye, thyroid inhibitor in the treatment of hyperthyroidism, as the acid tartrate and as a therapeutic drug for prion diseases. It is also used to kill bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, alcohol and ether.

Notes
Incompatible with strong oxidizing agents, strong acids, acid chlorides, acid anhydrides, nitric acid and sulfuric acid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-50-4
Molecular Formula C3H4N2S
Molecular Weight (g/mol) 100.14
MDL Number MFCD00005325
InChI Key RAIPHJJURHTUIC-UHFFFAOYSA-N
Synonym 2-aminothiazole, aminothiazole, 2-thiazolamine, thiazol-2-amine, abadol, 2-thiazolylamine, abadole, basedol, 2-thiazylamine, 4-thiazolin-2-onimine
PubChem CID 2155
ChEBI CHEBI:40782
IUPAC Name 1,3-thiazol-2-amine
SMILES NC1=NC=CS1

Specifications

Melting Point 88°C to 92°C
Boiling Point 117°C (15 mmHg)
Quantity 100 g
Beilstein 105738
Merck Index 14,479
Solubility Information Soluble in water,alcohol and ether.
Formula Weight 100.14
Percent Purity 97%
Chemical Name or Material 2-Aminothiazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Aminothiazole, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.