missing translation for 'onlineSavingsMsg'
Learn More
2-Amino-5-phenylpyrazine, 98%, Thermo Scientific™ Product Code.: 15276028

2-Amino-5-phenylpyrazine, 98%, Thermo Scientific™

Product Code. p-7077385 Shop All Thermo Scientific Chemicals Products
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15276028

Brand: Thermo Scientific Acros 452500010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 13535-13-2
Molecular Formula C10H9N3
Molecular Weight (g/mol) 171.20
MDL Number MFCD01646266
InChI Key KJAKXVBZQBPPOB-UHFFFAOYSA-N
Synonym 2-amino-5-phenylpyrazine, pyrazinamine, 5-phenyl, pyrazine, 2-amino-5-phenyl, acmc-1bpgx, 5-phenyl-2-pyrazinamine, 5-phenyl-2-aminopyrazine, 2-pyrazinamine,5-phenyl, 5-phenylpyrazine-2-ylamine, 5-phenyl-2-pyrazinamine #
PubChem CID 598734
IUPAC Name 5-phenylpyrazin-2-amine
SMILES NC1=NC=C(N=C1)C1=CC=CC=C1

Specifications

Melting Point 142.0°C to 148.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 171.2
Percent Purity 98%
Chemical Name or Material 2-Amino-5-phenylpyrazine
compliance-icons
Product Identifier
  • 2-Amino-5-phenylpyrazine, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Amino-5-phenylpyrazine, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.