missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-4-(4-bromophenyl)pyrimidine, 98%, Thermo Scientific™

Product Code. 15476387
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 392326-81-7
Molecular Formula C10H8BrN3
Molecular Weight (g/mol) 250.099
MDL Number MFCD02317272
InChI Key ULMOYCJVOJTPCJ-UHFFFAOYSA-N
Synonym 4-4-bromophenyl pyrimidin-2-amine, 2-amino-4-4-bromophenyl pyrimidine, 2-pyrimidinamine,4-4-bromophenyl, 4-4-bromophenyl-2-pyrimidinylamine, 2-pyrimidinamine, 4-4-bromophenyl, 4-4-bromo-phenyl-pyrimidin-2-ylamine
PubChem CID 4590150
IUPAC Name 4-(4-bromophenyl)pyrimidin-2-amine
SMILES C1=CC(=CC=C1C2=NC(=NC=C2)N)Br

Specifications

Melting Point 204°C to 207°C
Quantity 1 g
Formula Weight 250.1
Percent Purity 98%
Chemical Name or Material 2-Amino-4-(4-bromophenyl)pyrimidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.