missing translation for 'onlineSavingsMsg'
Learn More
2-Amino-2-ethyl-1,3-propanediol, 97% Product Code.: 10378310

2-Amino-2-ethyl-1,3-propanediol, 97%

Product Code. p-54730 Shop All Thermo Scientific Chemicals Products
500 g, Glass bottle, 500g
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
2.50kg
500g
This item is not returnable. View return policy

Product Code. 10378310

Brand: Thermo Scientific Acros 227115000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 115-70-8 | C5H13NO2 | 119.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-70-8
Molecular Formula C5H13NO2
Molecular Weight (g/mol) 119.16
MDL Number MFCD00004680
InChI Key IOAOAKDONABGPZ-UHFFFAOYSA-N
Synonym 2-amino-2-ethyl-1,3-propanediol, aepd, aminoethyl propanediol, 1,3-propanediol, 2-amino-2-ethyl, 2-ethyl-2-aminopropanediol, 2-amino-2-ethylpropanediol, unii-0adf1987d4, 2-amino-2-ethyl-propane-1,3-diol, 2-amino-1,3-dihydroxy-2-ethylpropane, aepd-85
PubChem CID 8282
IUPAC Name 2-amino-2-ethylpropane-1,3-diol
SMILES CCC(CO)(CO)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 35°C to 38°C
Color Yellow
Density 1.0990g/mL
Boiling Point 152°C to 153°C (10.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula HOCH2C(C2H5)(NH2)CH2OH
Refractive Index 1.4851 to 1.4871
Quantity 500 g
Merck Index 15, 436
Specific Gravity 1.099
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols
Formula Weight 119.16
Percent Purity 97%
Physical Form Viscous Liquid
Chemical Name or Material 2-Amino-2-ethyl-1, 3-propanediol, 97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0456856 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code227115000,227110025
Lot NumberA0465248 Certificate TypeCertificate of Analysis Date09/08/2024 Product Code227115000,227110025
Lot NumberA0456856 Certificate TypeCertificate of Analysis Date23/10/2023 Product Code227115000,227110025
Lot NumberA0435298 Certificate TypeCertificate of Analysis Date24/04/2023 Product Code227115000,227110025
Lot NumberA0421480 Certificate TypeCertificate of Analysis Date18/08/2020 Product Code227115000,227110025
compliance-icons
Product Identifier
  • 2-Amino-2-ethyl-1,3-propanediol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Amino-2-ethyl-1,3-propanediol, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.